Learn More
Apexbio Technology LLC DCEBIO 60563-36-2 10mg
SDP
Supplier:  Apexbio Technology LLC B673310
Encompass_Preferred
DCEBIO (CAS 60563-36-2) is a small molecule derivative of 1-EBIO recognized for its modulatory effects on ion channels In T84 colonic epithelial cells DCEBIO enhances chloride secretion by activating the human intermediate conductance Ca2 -activated K channel (hIK1) and facilitating apical membrane Cl- conductance thereby influencing epithelial electrolyte transport This compound is utilized in cell-based research to investigate ion channel-mediated signal transduction pathways Its ability to modulate specific ion channels supports its widespread use in studies focused on ion channel function and the modeling of disorders related to epithelial ion transport
Catalog No. 50-000-88845