Learn More
Medchemexpress LLC Dcemm2 | 296771-07-8 | 99.3% | 370.82 g/mol | C16H11ClN6OS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1449715MG
Encompass_Preferred
dCeMM2 is a molecular glue-type degrader that induces ubiquitination and proteasomal degradation of cyclin K by promoting interaction between CDK12-cyclin K and a CRL4B E3 ligase complex. It inhibits CDK12/13 activity and is intended for cell-based research applications.
  • Promotes ubiquitination and degradation of cyclin K in cells
  • Inhibits CDK12/13 enzymatic activity with selectivity over CDK7
  • Demonstrated cellular activity in KBM7 and HEK assays
  • High purity suitable for biochemical and cell assays
  • Soluble in DMSO for in vitro preparations
  • Stable as powder at -20°C or 4°C with defined storage times
Catalog No. 50-002-21307