Learn More
Medchemexpress LLC dCeMM2 | 296771-07-8 | 99.3% | C16H11ClN6OS | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14497150MG
Encompass_Preferred
dCeMM2 (Compound 2) is a molecular glue-type degrader that targets cyclin K. It induces ubiquitination and degradation of cyclin K by promoting an interaction of CDK12-cyclin K with a CRL4B ligase complex. It is for research use only and not sold to patients.
  • Molecular glue-type degrader
  • Targets cyclin K
  • Induces ubiquitination and degradation of cyclin K
  • Inhibits enzymatic activity of CDK12/13 with remarkable selectivity over CDK7
  • Promotes interaction between CDK12 and DDB1
  • For research use only
Catalog No. 50-003-55376