Learn More
AdipoGen DCIA
SDP
Supplier:  AdipoGen CDXD0090M020
Encompass_Preferred
Chemical. CAS 76877-34-4. Formula C22H23IN2O3. MW 490.33. Synthetic. A label that can be used for preparing maleimide and iodoacetamide derivatives of 3-phenylcoumarin fluorophore. These derivatives can be self-quenching until they react with thiol groups and regain their strong fluorescence.
Catalog No. 50-256-1247