Learn More
TARGETMOL CHEMICALS INC DCLK1-IN-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T84185MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. DCLK1-IN-1 is a selective and in vivo-compatible chemical probe of the DCLK1 kinase domain . purity: 98%
Catalog No. 50-257-9078