Learn More
Medchemexpress LLC Dclx069 | 792946-69-1 | 98.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY122096100MG
Encompass_Preferred
DCLX069 is a selective protein arginine methyltransferase 1 (PRMT1) inhibitor with an in vitro IC50 of 17.9 μM that shows reduced activity against PRMT4 and PRMT6 and has demonstrated antiproliferative effects in cancer cell lines.
  • Selective PRMT1 inhibition with IC50 17.9 μM.
  • Less active against PRMT4 and PRMT6, indicating selectivity.
  • Demonstrated anticancer activity in in vitro cell proliferation assays.
  • Molecular formula C20H25N3O2 and molecular weight 339.43.
  • Available in small-molecule research pack sizes and as a ready-to-use DMSO solution for reconstitution.
Catalog No. 50-002-30845