Learn More
Medchemexpress LLC Dclx069 | 792946-69-1 | 98.0% | 50 MG

Supplier: Medchemexpress LLC HY12209650MG
DCLX069 is a selective small-molecule inhibitor of protein arginine methyltransferase 1 (PRMT1) with reported anticancer activity; it inhibits PRMT1 with an IC50 of 17.9 μM and shows reduced activity against PRMT4 and PRMT6. Supplied as a light yellow solid for research use.
- Selective PRMT1 inhibitor with IC50 17.9 μM.
- Shows lower activity against PRMT4 and PRMT6.
- Molecular weight 339.43 g/mol; formula C20H25N3O2.
- High purity (98.0%) and light yellow solid appearance.
- Store at 4 °C and protect from light; in solution store at -80 °C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.