Learn More
Medchemexpress LLC DD-03-156 (S,R,S)-AHPC-Me-PEG2-dabrafenib | 2769753-69-5 | 99.9% | 1106.31 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13734650MG
Encompass_Preferred
DD-03-156 is a potent and selective degrader of CDK17 and LIMK2. Its selectivity and potency make it an advanced starting point for developing a chemical probe for the degradation of CDK17.
  • Potent and selective degrader of CDK17 and LIMK2.
  • Advanced starting point for developing a chemical probe for CDK17 degradation.
  • Involves a VHL ligand, a linker, and a BRAF inhibitor.
  • For research use only.
Catalog No. 50-003-49629