Learn More
STA PHARMACEUTICAL US LLC Dde-L-Orn(Fmoc)-OH | 5 g | CAS 1423017-87-1 | MDL MFCD06796885
Supplier: STA PHARMACEUTICAL US LLC STAAA113495GM
Dde-L-Orn(Fmoc)-OH is a Amino Acid reagent (Subcategory: Lys) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1423017-87-1
- MDL: MFCD06796885
- InChIKey: GHZOEWCLMLYIMI-DEOSSOPVSA-N
- Molecular Weight: 518.61
- Molecular Formula: C30H34N2O6
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S)-5-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-((1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethyl)amino)pentanoic acid
- SMILES: O=C(OCC1C2=CC=CC=C2C3=C1C=CC=C3)NCCC[C@@H](C(O)=O)N/C(C)=C4C(CC(C)(C)CC\4=O)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.