Learn More
American Radiolabeled Chemicals Inc DDG 14C(U) C 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ARC 0112C50UC
Encompass_Preferred
Deoxy-D-glucose, 2-14C(U) 50uCi in sterile saline, 0.1 mCi/ml, 300-360 mCi/mmol, M.W. 164.2
Catalog No. 50-352-927