Learn More
Medchemexpress LLC DDR1-IN-4 1mg | 2125676-13-1 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1141731MG
Encompass_Preferred
DDR1-IN-4 is a selective small-molecule inhibitor of Discoidin Domain Receptor 1 (DDR1) that inhibits DDR1 autophosphorylation (IC50 29 nM) and shows reduced activity against DDR2 (IC50 1.9 μM). It is supplied as a high-purity research reagent (CAS 2125676-13-1) with solubility data supporting in vitro and in vivo formulation.
  • Selective inhibition of DDR1 (IC50 29 nM).
  • Lower potency against DDR2 (IC50 1.9 μM).
  • High purity for reliable experimental results.
  • High solubility in DMSO (250 mg/mL) for in vitro use.
  • Offered in milligram-scale packages for research applications.
Catalog No. 50-003-07552