Learn More
Medchemexpress LLC DDR1-IN-9 | 1934246-20-4 | >98% | 584.66 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1677455MG
Encompass_Preferred
DDR1-IN-9 is a potent, selective, and orally bioavailable inhibitor of discoidin domain receptor 1 (DDR1) with an IC50 of 0.4 nM. It shows >1000-fold selectivity over DDR2 and other kinases. DDR1-IN-9 can be used for research on chronic kidney disease and fibrosis.
  • Inhibits discoidin domain receptor 1 (DDR1) with high potency.
  • Demonstrates excellent selectivity for DDR1 over DDR2 and other kinases.
  • Orally bioavailable compound.
  • Potentially useful in chronic kidney disease research.
  • Potentially useful in fibrosis research.
Catalog No. 50-003-90576