Learn More
Aviva Systems Biology Corp INTS6L Antibody - N-terminal region (ARP36539_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36539_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DDX26B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DDX26B. Alternate names: DKFZp686G0470, FLJ41215, MGC88298ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56183