Learn More
Cayman Chemical De-Boc-CabazItaxel 1mg
SDP
Supplier:  Cayman Chemical 401161 MG
Encompass_Preferred
A potential impurity in commercial preparations of cabazitaxel; has been used in the synthesis of tubulin-targeting fluorescent dyes
Catalog No. 50-294-4054