Learn More
Cayman Chemical DEA NONOate, 372965-00-9, 50 mg
SDP
Supplier:  Cayman Chemical 8210050
Encompass_Preferred
Molecular Formula C4H11N3O2 • C4H11N, Purity ≥98%, Formula Weight 206.3, Synonyms: Diethylammonium (Z)-1-(N,N-diethylamino)diazen-1-ium-1,2-diolate, Diethylamine NONOate, DEA/NO; A NO donor with a half-life of 2 minutes at 37C, pH 7.4; liberates 1.5 moles of NO per mole of parent compound.
Catalog No. NC0955840
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.