Learn More
Medchemexpress LLC Deacetyl Ganoderic Acid F | 100665-44-9 | 1 MG

Supplier: Medchemexpress LLC HYN35011MG
Deacetyl Ganoderic Acid F is a 7-anostane triterpenoid from *Ganoderma lucidum*. It is presented as a white to off-white solid with a purity of 98.82%.
- Purity of 98.82%
- Molecular weight: 528.63
- Formula: C30H40O8
- Appearance: White to off-white solid
- Structure classification: Terpenoids, triterpenes
- Initial source: Plants, Polyporaceae
- For research use only
- Store at 4°C, sealed, away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.