Learn More
Bachem (Asp28)-Glucagon (1-29) (human, rat, porcine) Trifluoroacetate
SDP
Supplier:  Bachem 4079056.1000
Encompass_Preferred
(Asp28)-Glucagon (1-29) (human, rat, porcine) Trifluoroacetate, H-His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asp-Thr-OH, 1037751-81-7, C153H224N42O50S, Mr 3483.78, 1mg. Deamidation product of glucagon.
Catalog No. 50-194-6897