Learn More
Medchemexpress LLC Decabromodiphenyl ethane | 84852-53-9 | 98.0% | 971.22 | 500 G
Supplier: Medchemexpress LLC HYW013058500G
Decabromodiphenyl ethane (DBDPE; 1,2-Bis(perbromophenyl)ethane) is a brominated flame retardant. It has been observed to exhibit neurotoxicity, thyroid toxicity, reproductive developmental toxicity, hepatotoxicity, and oxidative stress in living organisms.
- Brominated flame retardant
- Can exhibit neurotoxicity
- Can exhibit thyroid toxicity
- Can exhibit reproductive developmental toxicity
- Can exhibit hepatotoxicity
- Can exhibit oxidative stress in living organisms