Learn More
Electron Microscopy Sciences Hard Specimen Decalcifier 1 LT
SDP
Supplier:  Electron Microscopy Sciences 6414701
Encompass_Preferred
Ready-to-use decalcifier is ideal for hard specimens and protects the cellular structure with minial interference with subsequent staining. For use with human and animal calcified tissue, bone, cartilage and other hard specimens.
Catalog No. 50-252-93