Learn More
Medchemexpress LLC Decanoic acid, 4-(4-chlorophenyl)-1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidinyl ester | 74050-97-8 | MFCD00866706 | 99.7% | 530.11 g/mol | C31H41ClFNO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1079695MG
Encompass_Preferred
Haloperidol decanoate is the decanoate ester of haloperidol, a butyrophenone-class antipsychotic. It is a long-acting, lipophilic ester used in research to model depot formulations and to study dopamine D2 receptor pharmacology. Supplied as a solid for analytical and preclinical use; consult the batch certificate of analysis for purity and storage instructions.
  • Long-acting decanoate ester for depot formulation studies.
  • Dopamine D2 receptor antagonist activity relevant to antipsychotic research.
  • High purity suitable for analytical and preclinical applications.
  • Available in small research quantities for dosing and formulation work.
  • Refer to batch certificate of analysis for purity, identity, and storage details.
Catalog No. 50-002-42949