Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-2-hydroxyglutaric acid disodium | 63512-50-5 | MFCD00021700 | 99.1% | 192.08 g/mol | C5H6Na2O5 | 100 MG
SDP

Supplier:  Medchemexpress LLC HYW015114100MG

Encompass_Preferred

L-2-Hydroxyglutaric acid disodium is the disodium salt of L-2-hydroxyglutaric acid used as a research reagent in epigenetic and metabolic studies. It acts as a putative oncometabolite that can inhibit histone demethylases and alter histone methylation, and it has been reported to inhibit mitochondrial creatine kinase activity.

  • High purity suitable for research use.
  • Molecular weight 192.08 g/mol.
  • Chemical formula C5H6Na2O5.
  • Available in small milligram pack sizes, including 100 mg.
  • Intended for biochemical assays and epigenetic studies.
  • Product documentation includes SDS and data sheet for handling and safety.

Catalog No. 50-003-02594


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.