Learn More
Medchemexpress LLC Octanoic acid, 4-ethyl- | 16493-80-4 | MFCD00506494 | 99.1% | 172.27 g/mol | C10H20O2 | 1 G

Supplier: Medchemexpress LLC HYW0153071G
4-Ethyloctanoic acid (octanoic acid, 4-ethyl-) is a medium-chain branched fatty acid reported as a natural compound first isolated from Saussurea lappa Clarke. It is used in flavor and food additive studies and is supplied for research, analytical standards, and formulation work as a high-purity reagent suitable for laboratory use.
- Medium-chain branched fatty acid (C10H20O2).
- High purity (≈99.1%).
- Molecular weight 172.27 g/mol.
- Suitable for flavor and food additive research and formulation studies.
- Documentation available: COA and SDS.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.