Learn More
Medchemexpress LLC Decanoic acid-d5 | 1219803-00-5 | 98.3% | 177.30 | C10H15D5O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW015309S3100MG
Encompass_Preferred
Decanoic acid-d5 is the deuterium-labeled analogue of decanoic (capric) acid, provided as an oil for use as an isotope-labeled standard or tracer in analytical and research workflows. It is reported at 98.3% purity with a molecular weight of 177.30, and is intended for quantitative assays, method development, and spectroscopic studies.
Catalog No. 50-003-02627