Learn More
American Radiolabeled Chemicals Inc DECANOYL 3H CO A 50UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 191150UC
Encompass_Preferred
n-Decanoyl-2,3-3H Coenzyme A 50uCi in ethanol:0.01N NaOAc (pH 5.2) (1:1), 1 mCi/ml, 50-60 Ci/mmol, M.W. 921.8, Shipped in Dry Ice, Exclusive
Catalog No. 50-352-926
Request a Quote
This product is regulated by the federal government, and may not be ordered online.