Learn More
Medchemexpress LLC Decanoyl-L-carnitine | 3992-45-8 | 98.0% | 315.45 g/mol | C17H33NO4 | 5 MG

Supplier: Medchemexpress LLC HY1350355MG
Decanoyl-L-carnitine is an acylcarnitine derivative used as a research reagent and diagnostic metabolite biomarker. It stimulates formation of desaturated fatty acid metabolites from [1-14C]-22:4 (n-6) and [1-14C]-22:5 (n-3). For research use only.
- Used as a diagnostic metabolite biomarker and research reagent.
- Stimulatory effect on formation of desaturated fatty acid metabolites.
- High purity: 98.0%.
- Solid, white to off-white appearance; molecular weight 315.45 g/mol.
- Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
- Powder storage stable at -20°C for up to 3 years.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.