Learn More
Medchemexpress LLC Decanoyl-L-carnitine-d3 (chloride) | 2483831-87-2 | MFCD08705111 | 99.9% | 354.93 | C17H31D3ClNO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY132949S5MG
Encompass_Preferred
Decanoyl-L-carnitine-d3 (chloride) is the deuterium-labeled chloride salt of decanoyl-L-carnitine, supplied as an isotopically labeled reagent for use as an internal standard in quantitative mass spectrometry. It is intended for research use and is characterized by high isotopic enrichment and analytical-grade purity suitable for LC-MS and GC-MS workflows.
  • Deuterium-labeled internal standard for accurate quantification by LC-MS and GC-MS.
  • High chemical purity (>99.9%) for reliable analytical performance.
  • Provided as a stable solid, suitable for long-term storage.
  • Chloride salt form compatible with common analytical protocols.
  • Three deuterium atoms provide a clear mass shift from the native analyte.
Catalog No. 50-002-62497