Learn More
Medchemexpress LLC Decanoyl-RVKR-CMK | 150113-99-8 | MFCD00798792 | 98.0% | 744.41 | C34H66ClN11O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10776010MG
Encompass_Preferred
Decanoyl-RVKR-CMK is a peptidic inhibitor of subtilisin/Kex2p-like proprotein convertases, including furin. It blocks envelope glycoprotein precursor processing and gp160 maturation, and has been used to inhibit HIV-1 and HIV-2 replication in cell models. Typical applications include studies of viral glycoprotein processing, secretory pathway regulation, and protease biology.
  • Inhibits furin and related proprotein convertases.
  • Blocks envelope glycoprotein precursor processing and gp160 maturation.
  • Used to probe viral replication and glycoprotein processing in cell models.
  • Supplied as a purified research reagent (purity 98.0%).
  • Molecular weight 744.41; formula C34H66ClN11O5.
Catalog No. 50-002-41198