Learn More
Medchemexpress LLC Decapeptide-12 | 137665-91-9 | 99.9% | 1395.52 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP009610MG
Encompass_Preferred
Decapeptide-12 is a small oligopeptide and a tyrosinase inhibitor that interacts with the C-terminal residue of tyrosinase (Kd: 61.1 μM). It is a competitive inhibitor of mushroom tyrosinase (IC50: 40 μM) and also increases the transcription of SIRT. It reduces melanin content in melanocytes and is used for research in melanogenesis, senescence, and inflammation.
  • Tyrosinase inhibitor
  • Reduces melanin content in melanocytes
  • Increases transcription of SIRT
  • Used for research of melanogenesis, senescence, and inflammation
  • Soluble in H2O (≥ 100 mg/mL or 71.66 mM)
Catalog No. 50-004-26030