Learn More
Medchemexpress LLC Dechloro Rivaroxaban | 1415566-28-7 | MFCD31654721 | 99.7% | C19H19N3O5S | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY76971
Encompass_Preferred
Dechloro Rivaroxaban is a highly selective, orally active inhibitor of Factor Xa. It effectively inhibits human free FXa with a Ki of 0.4 nM and demonstrates inhibition of prothrombinase activity and fibrin-associated FXa activity with IC50s of 2.1 nM and 92 nM, respectively.
  • Highly selective Factor Xa inhibitor
  • Orally active compound
  • Inhibits human free FXa with a Ki of 0.4 nM
  • Inhibits prothrombinase activity with an IC50 of 2.1 nM
  • Inhibits fibrin-associated FXa activity with an IC50 of 92 nM
  • Appears as a white to off-white solid
Catalog No. 50-003-58484