Learn More
AdipoGen Decitabine
SDP
Supplier:  AdipoGen CDXA0563M010
Encompass_Preferred
Chemical. CAS 2353-33-5. Formula C8H12N4O4. MW 228.21. Decitabine is an epigenetic modifier that inhibits DNA methyltransferase activity which results in DNA demethylation hypomethylation and gene activation by remodeling "opening" chromatin. This cytosine analog incorporates into DNA, inhibit DNA methyltransferases, and cause hypomethylation of cytosine residues in the absence of any significant mutagenic effects. Decitabine is a potent antileukaemic agent with activity in the treatment of haematological malignancies such as acute myeloid leukaemia AML, chronic myeloid leukaemia CML and myelodysplastic syndrome preleukaemia and has been suggested to reverse chemotherapy resistance in several cancer types. It is used as a chemotherapeutic agent, suppresses growth of human tumor cell lines, shown to reverse embryonic stem cell differentiation and prevent atherosclerosis lesion formation and reduces the production of inflammatory cytokines by macrophages.
Catalog No. 50-256-1735