Learn More
Medchemexpress LLC Decitabine | 2353-33-5 | 99.9% | C8H12N4O4 | 2 G
SDP
Supplier:  Medchemexpress LLC HYA00042G
Encompass_Preferred
Decitabine is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. It incorporates into DNA in place of cytosine, which can covalently trap DNA methyltransferase to DNA, leading to irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis, demonstrating potent anticancer activity.
  • Deoxycytidine analogue antimetabolite
  • DNA methyltransferase inhibitor
  • Induces cell G2/M arrest
  • Induces cell apoptosis
  • Exhibits potent anticancer activity
Catalog No. 50-004-33071