Learn More
TARGETMOL CHEMICALS INC Decitabine 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1508500MG
Encompass_Preferred
Also available in 1 g, 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Decitabine (Deoxycytidine) is a deoxycytidine analog, a DNA methyltransferase inhibitor with oral activity. Decitabine has antitumor activity and antimetabolic activity. Decitabine induces cell cycle arrest and apoptosis. Purity 99.17%
Catalog No. 50-287-6691