Learn More
Medchemexpress LLC Decitabine (Standard) | 2353-33-5 | 99.9% | C8H12N4O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYA0004R
Encompass_Preferred
Decitabine (Standard) is the analytical standard of Decitabine, intended for research and analytical applications. It is an orally active deoxycytidine analogue antimetabolite and a DNA methyltransferase inhibitor. It incorporates into DNA in place of cytosine, covalently trapping DNA methyltransferase and causing irreversible inhibition of the enzyme. Decitabine induces cell G2/M arrest and cell apoptosis and exhibits potent anticancer activity.
  • Analytical standard for research and analytical applications
  • Orally active deoxycytidine analogue antimetabolite
  • DNA methyltransferase inhibitor
  • Induces cell G2/M arrest and cell apoptosis
  • Exhibits potent anticancer activity
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments
Catalog No. 50-003-57849