Learn More
Medchemexpress LLC Decloxizine dihydrochloride | 13073-96-6 | MFCD04972004 | 99.7% | 413.38 g/mol | C21H30Cl2N2O2 | 10 MG

Supplier: Medchemexpress LLC HYA007510MG
Decloxizine dihydrochloride is the dihydrochloride salt of decloxizine, a histamine H1 receptor antagonist supplied as a research reagent. The material is intended for laboratory research use only and is not for human or clinical use.
- For research use only; not for human or clinical application.
- High purity (≈99.7%).
- Molecular weight 413.38 g/mol.
- Molecular formula C21H30Cl2N2O2.
- CAS number 13073-96-6.
- Supplied as a 10 MG solid; store sealed at 4°C, away from moisture.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.