Learn More
Cayman Chemical Decnoyl-L-carnItInhydrate 10mg
SDP
Supplier:  Cayman Chemical 2690910 MG
Encompass_Preferred
A derivative of L-carnitine; increases the formation of C24 fatty acid intermediates, as well as docosapentaenoic and docosahexaenoic acid in rat hepatocytes
Catalog No. 50-293-7111