Learn More
ELITechGroup Aerospray Gram Reagent A, Decolorizer with Safranine, 500mL
SDP
Supplier:  ELITechGroup SS041A
Encompass
This item has a minimum order quantity of 2 per supplier requirements. Gram Reagent A, Decolorizer with Safranin,500 ml,Chemical Name:Alcohol Based Gram Stain,Flash Point:11degreeC,Apperance:Liquid,Odor:Characteristic Alcohol, color:Red
Catalog No. NC9419518