Learn More
Apexbio Technology LLC Decoquinate 18507-89-6 100mg
SDP
Supplier:  Apexbio Technology LLC B1722100
Encompass_Preferred
Decoquinate (CAS 18507-89-6) is a small-molecule inhibitor targeting the mitochondrial electron transport chain in coccidian parasites It is designed to interfere with mitochondrial bioenergetics thereby suppressing cellular energy metabolism and parasite proliferation Decoquinate exerts its biological activity primarily through disruption of electron transport chain activity In in vitro studies Decoquinate demonstrates potent inhibition with reported IC50 values in the nanomolar range against Plasmodium falciparum cultured cells Based on these pharmacological properties Decoquinate holds research potential in studying mitochondrial metabolism in protozoan parasites investigating host-parasite interactions and exploring antimalarial drug development
Catalog No. 50-000-96491