Learn More
Medchemexpress LLC Decoquinate | 18507-89-6 | 99.3% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB10361G
Encompass_Preferred
Decoquinate is an orally active, selective inhibitor of the mitochondrial bc1 complex, primarily used in veterinary research. It targets Eimeria spp. sporozoites and first generation schizonts, as well as Plasmodium spp. By inhibiting electron transfer through competitive binding to the mitochondrial cytochrome b system, it blocks the parasite's energy metabolism, thereby inhibiting its development and reproduction.
  • Targets Eimeria spp. and Plasmodium spp.
  • Inhibits mitochondrial bc1 complex and electron transfer
  • Blocks parasite energy metabolism
  • Exhibits significant anticoccidial activity
  • Prevents intestinal damage
  • Improves host growth performance
  • Has inhibitory effects on liver and blood stages of Plasmodium
  • Used for prevention and treatment of coccidiosis in ruminants and poultry
Catalog No. 50-004-13795