Learn More
Electron Microscopy Sciences Serum Vial De-Crimpier Pliers for sizes 8 and 30 mm
SDP
Supplier:  Electron Microscopy Sciences 6218002
Encompass_Preferred
Made from stainless steel offers one tool for both sizes: 13 mm and 20 mm serum vial de-crimpier. An inexpensive and easy way to remove the seal from the serum vial, simply secure the seal between the jaws, squeeze the handle and twist the seal. Plastic coated handle offers comfort and ease of use.
Use: 8 and 30 mm
Catalog No. 50-327-81