Learn More
Medchemexpress LLC Dectin-1/CLEC7A protein, human, Fc fusion | >95.0% | 100UG
SDP
Supplier:  Medchemexpress LLC HYP70101100UG
Encompass_Preferred
Recombinant human Dectin-1/CLEC7A Fc fusion protein containing the C-type lectin domain. Produced in HEK293 cells and supplied purified for use in ligand binding studies, receptor characterization, and assay development. The Fc fusion increases stability and enables easy detection or immobilization in immunoassays.
Catalog No. 50-001-75401