Learn More
Cayman Chemical DecursInol ngelate 10mg
SDP
Supplier:  Cayman Chemical 2521210 MG
Encompass_Preferred
A pyranocoumarin with diverse biological activities; inhibits the growth of various cancer cell lines (ED50s = 24-56 μM); decreases estrogen-stimulated growth of MCF-7 breast cancer cells; activates PKC purified from rat brain; selectively inhibits CYP1A1/2 over CYP2D15 and CYP3A12 in canine liver microsomes (Kis = 67.56, 872.5, and 853.9 μM, respectively); reduces Aβ25-35-induced cytotoxicity and MDA production and increases SOD and GPx activities in PC12 cells; has antinociceptive activity in the tail flick and hot plate tests and reduces acetic acid-induced writhing in mice at 200 mg/kg
Catalog No. 50-293-3866