Learn More
Medchemexpress LLC Decyl glucoside | 68515-73-1 | MFCD00063297 | 60.0% | 320.42 g/mol | C16H32O6 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW25017010G
Encompass_Preferred
Decyl glucoside (60% in water) is a mild, biodegradable nonionic surfactant supplied as an aqueous solution for laboratory and formulation use. Derived from glucose and fatty alcohols, it functions as a gentle detergent and emulsifier for research and small-scale formulation work. The product is a colorless to light yellow liquid and is stored under recommended conditions for stability.
  • Mild, nonionic surfactant suitable for sensitive formulations.
  • Biodegradable and derived from renewable sources.
  • Aqueous 60% formulation for easier handling and dilution.
  • Liquid form, colorless to light yellow, ready for formulation.
  • Useful as a detergent, emulsifier, and solubilizer in research applications.
  • Stable when stored at recommended conditions (store at -20°C).
Catalog No. 50-002-82571