Learn More
Medchemexpress LLC Decyl glucoside | 68515-73-1 | MFCD00063297 | 60.0% | 320.42 g/mol | C16H32O6 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW2501705G
Encompass_Preferred
Decyl glucoside is a mild, nonionic surfactant supplied as a 60% aqueous solution. Derived from glucose and fatty alcohols, it is used for foaming, cleansing, and formulation research. The liquid reagent offers good biodegradability and compatibility with sensitive formulations; follow the product SDS for handling and storage.
  • Mild nonionic surfactant suitable for gentle formulations.
  • Excellent foaming and cleaning properties.
  • Derived from renewable glucose and fatty alcohols.
  • Supplied as a 60% solution in water for ready use.
  • Colorless to light yellow liquid appearance.
  • Biodegradable and formulation-friendly.
  • Intended for research use only; follow SDS for storage and handling.
Catalog No. 50-002-82250