Learn More
Medchemexpress LLC Decyltrimethylammonium bromide | 2082-84-0 | MFCD00041973 | 98.0% | 280.29 | 25 G
Supplier: Medchemexpress LLC 25GHYW008630
Decyltrimethylammonium bromide is a quaternary ammonium salt comprising a decyltrimethylammonium cation and a bromide anion. This biochemical reagent is intended for life science-related research as a biological material or organic compound, with a role as a surfactant. It is for research use only and not sold to patients.
- Solid form
- White to light yellow color