Learn More
Analytical Sales & Services Deep Square Well Plate for KingFisher™ Extraction and Purification Systems
Supplier:  Analytical Sales & Services 969013
Encompass
-For use with KingFisher™ Duo Prime, Flex, Apex, and Presto. -Alphanumeric well coding. -Available sterile or non-sterile. -Standardized plate geometry (conforms to ANSI 1-2004. -Length x Width: 127.76mm x 85.48mm. -Free of detectable DNase, RNase. -Non-pyrogenic. -Manufactured under ISO 9001 guidelines. -Ideal microplates for magnetic particle processing applications. -Compatible with KingFisher™ automated systems. -Uniform external dimensions and tolerances. -Solid bottom. -Well profile: V-bottom (conical bottom). -Volume: 2.2mL.
Catalog No. NC3735621