Learn More
Analytical Sales & Services Deep Well Reservoir, Two Columns
Supplier:  Analytical Sales & Services 962144
Encompass
Clear Polypropylene Deep Well Reservoir, Two Columns, 144mL per Column, 285mL maximum. Height: 44mm, Well Bottom Shape: Pyramid
Catalog No. NC1247565