Learn More
Medchemexpress LLC Defactinib hydrochloride | 1073160-26-5 | 99.4% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY12289A100MG
Encompass_Preferred
Defactinib hydrochloride is the hydrochloride salt of defactinib, a small-molecule focal adhesion kinase (FAK) inhibitor that reduces FAK phosphorylation at Tyr397 in a time- and dose-dependent manner. It is used in preclinical research to study FAK signaling, cancer cell migration, and related pathways.
  • Small-molecule FAK inhibitor reducing phosphorylation at Tyr397.
  • Used for preclinical studies of cancer cell migration and signaling.
  • High purity suitable for biochemical and cellular assays.
  • Solid form, typically supplied as a 100 mg package.
  • Store sealed at 4°C; in solution store at -80°C for long-term storage.
Catalog No. 50-002-30352