Learn More
Selleck Chemical LLC Defensamide
SDP
Supplier:  Selleck Chemical LLC S65122MG
Encompass_Preferred
Defensamide (MHP) is an activator of sphingosine kinase (SPHK1) It can modulate the innate epidermal immune response by potentiating SPHK1 activity and inducing cAMP production
Catalog No. 50-000-04614