Learn More
Medchemexpress LLC Deferiprone | 30652-11-0 | MFCD00134497 | 100.0% | 139.15 g/mol | C7H9NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0568R10MG
Encompass_Preferred
Deferiprone is a small-molecule iron chelator supplied as an analytical reference standard for laboratory research. This solid reference material (10 mg) is intended for identity confirmation, method development, and quality control in chromatographic and mass spectrometry workflows.
  • High purity (99.99%) analytical reference standard.
  • Molecular weight 139.15 g/mol; formula C7H9NO2.
  • Suitable for HPLC, LC-MS, and identity confirmation.
  • Supplied as a solid 10 MG pack for easy handling and weighing.
  • Certificate of analysis and handling instructions available.
Catalog No. 50-002-98293