Learn More
TARGETMOL CHEMICALS INC DEFERITAZOLE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T313625MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Deferitazole (FBS 0701) is a novel and orally active selective and high affinity iron chelator. purity: 98%
Catalog No. 50-258-0370